Assay Details
| Gene Symbol: Gmps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033300.2 | NP_001028472.2 | 5 - 6 | 667 | 92 | |
| GenBank mRNA | AK049755.1 | - | 5 - 6 | 631 | 92 | |
| AK143197.1 | - | 5 - 6 | 668 | 92 | ||
| AK146654.1 | - | 5 - 6 | 685 | 92 | ||
| AK167857.1 | - | 5 - 6 | 647 | 92 | ||
| AK168239.1 | - | 5 - 6 | 617 | 92 | ||
| AK169043.1 | - | 5 - 6 | 651 | 92 | ||
| AK169701.1 | - | 5 - 6 | 665 | 92 | ||
| BC037232.1 | - | 3 - 4 | 319 | 92 | ||
| BC138411.1 | - | 5 - 6 | 623 | 8861431 | 92 | |
| BC138412.1 | - | 5 - 6 | 623 | 8861432 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmps
Genomic Map