Assay Details
| Gene Symbol: Gmps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033300.2 | NP_001028472.2 | 6 - 7 | 866 | 113 | |
| GenBank mRNA | AK049755.1 | - | 6 - 7 | 830 | 113 | |
| AK143197.1 | - | 6 - 7 | 867 | 113 | ||
| AK146654.1 | - | 6 - 7 | 884 | 113 | ||
| AK167857.1 | - | 6 - 7 | 846 | 113 | ||
| AK168239.1 | - | 6 - 7 | 816 | 113 | ||
| AK169043.1 | - | 6 - 7 | 850 | 113 | ||
| AK169701.1 | - | 6 - 7 | 864 | 113 | ||
| BC037232.1 | - | 4 - 5 | 518 | 113 | ||
| BC080685.1 | - | 1 - 2 | 78 | 113 | ||
| BC138411.1 | - | 6 - 7 | 822 | 8861431 | 113 | |
| BC138412.1 | - | 6 - 7 | 822 | 8861432 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmps
Genomic Map