Assay Details
| Gene Symbol: Gmps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033300.2 | NP_001028472.2 | 8 - 9 | 1182 | 91 | |
| GenBank mRNA | AK049755.1 | - | 8 - 9 | 1146 | 91 | |
| AK143197.1 | - | 8 - 9 | 1183 | 91 | ||
| AK146654.1 | - | 8 - 9 | 1200 | 91 | ||
| AK169043.1 | - | 8 - 9 | 1166 | 91 | ||
| AK169701.1 | - | 8 - 9 | 1180 | 91 | ||
| BC037232.1 | - | 6 - 7 | 834 | 91 | ||
| BC080685.1 | - | 3 - 4 | 394 | 91 | ||
| BC138411.1 | - | 8 - 9 | 1138 | 8861431 | 91 | |
| BC138412.1 | - | 8 - 9 | 1138 | 8861432 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmps
Genomic Map