Assay Details
| Gene Symbol: Gmps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033300.2 | NP_001028472.2 | 9 - 10 | 1354 | 104 | |
| GenBank mRNA | AK049755.1 | - | 9 - 10 | 1318 | 104 | |
| AK143197.1 | - | 9 - 10 | 1355 | 104 | ||
| AK146654.1 | - | 9 - 10 | 1372 | 104 | ||
| AK167857.1 | - | 9 - 10 | 1331 | 104 | ||
| AK168239.1 | - | 9 - 10 | 1304 | 104 | ||
| AK169043.1 | - | 9 - 10 | 1338 | 104 | ||
| AK169701.1 | - | 9 - 10 | 1352 | 104 | ||
| BC037232.1 | - | 7 - 8 | 1006 | 104 | ||
| BC080685.1 | - | 4 - 5 | 566 | 104 | ||
| BC138411.1 | - | 9 - 10 | 1310 | 8861431 | 104 | |
| BC138412.1 | - | 9 - 10 | 1310 | 8861432 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmps
Genomic Map