Assay Details
| Gene Symbol: Magea10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085506.1 | NP_001078975.1 | 4 | 1440 | 119 | |
| GenBank mRNA | AK136064.1 | - | 4 | 1441 | 119 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131213 | AK136064.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2131213 | NM_001085506.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116960 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179414 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131213 | AK136064.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2131213 | NM_001085506.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179414 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32457795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15046008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15046006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15056659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15056675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14787512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15056656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15211748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14787526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15056691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15056663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14787510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32457773 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10604959 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10604958 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 885461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216864 | - | - |

Magea10
Genomic Map