Assay Details
| Gene Symbol: Pnpla8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026164.2 | NP_080440.2 | 7 - 8 | 1908 | 89 | |
| GenBank mRNA | AB044139.1 | - | 7 - 8 | 1787 | 89 | |
| AK004621.1 | - | 7 - 8 | 1875 | 89 | ||
| AK084928.1 | - | 1575 | 89 | |||
| AK145776.1 | - | 7 - 8 | 1818 | 89 | ||
| AK163211.1 | - | 7 - 8 | 1876 | 89 | ||
| AK168475.1 | - | 7 - 8 | 1871 | 89 | ||
| BC019364.1 | - | 4 - 5 | 362 | 89 | ||
| BC127056.1 | - | 6 - 7 | 1768 | 40109753 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnpla8
Genomic Map