Assay Details
| Gene Symbol: Hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013920.2 | NP_038948.1 | 3 - 4 | 571 | 106 | |
| GenBank mRNA | AJ242626.1 | - | 3 - 4 | 523 | 106 | |
| BC125519.1 | - | 3 - 4 | 393 | 40130034 | 106 | |
More Information
Set Membership: |
|

Hnf4g
Genomic Map