Assay Details
| Gene Symbol: 4930568G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040480.1 | 2 - 3 | 147 | 111 | ||
| GenBank mRNA | AK016251.1 | - | 2 - 3 | 148 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700029M03 gene | 1700029M03Rik | -107.1kb | Anti-sense | Chr. 1 - 164960664 - 164961400 on NCBI Build GRCm38 | |
| Downstream | T-box 19 | Tbx19 |
AU019250, D1Ertd754e, Tpit |
68.4kb | Sense | Chr. 1 - 165137834 - 165160773 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930568G15Rik
Genomic Map