Assay Details
| Gene Symbol: 4930568G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040480.1 | 4 - 5 | 456 | 107 | ||
| GenBank mRNA | AK016251.1 | - | 4 - 5 | 457 | 107 | |
| AK036349.1 | - | 84 | 107 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700029M03 gene | 1700029M03Rik | -105.2kb | Anti-sense | Chr. 1 - 164960664 - 164961400 on NCBI Build GRCm38 | |
| Downstream | T-box 19 | Tbx19 |
AU019250, D1Ertd754e, Tpit |
69.7kb | Sense | Chr. 1 - 165137834 - 165160773 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2362645 | AK036349.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723911 | AK035496.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723911 | AK035765.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195334 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672715 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13099 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135311 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00239973 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135311 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00946947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00293344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00306044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00563189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00685286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00230677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00685284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00079586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00520788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00293343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00654240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01004176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00124027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00135364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00590562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00563193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01004174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01085717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01144319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00293339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00293328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00306040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00520801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00710948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00520792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00293332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00293345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01004178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00230670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01004173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20792763 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1430884_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430884_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430884_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100028174.mm.1 | NR_040480.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028174.mm.1 | NR_040480.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028174.mm.1 | TC0100003436.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430884_at | - | - |
| Mouse Expression Set 430 | 1430884_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430884_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430884_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 512057 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102450711 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102450711 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227170 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102450711 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102450711 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227170 | - | - |

4930568G15Rik
Genomic Map