Assay Details
| Gene Symbol: Lrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081406.1 | NP_001074875.1 | 1 - 2 | 251 | 73 | |
| GenBank mRNA | AK010419.1 | - | 1 - 2 | 244 | 73 | |
| AK144741.1 | - | 1 - 2 | 252 | 73 | ||
| BC022648.1 | - | 1 - 2 | 205 | 73 | ||
| BC075711.1 | - | 1 - 2 | 233 | 73 | ||
| BC117053.1 | - | 1 - 2 | 230 | 73 | ||
| BC117055.1 | - | 1 - 2 | 230 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrr1
Genomic Map