Assay Details
| Gene Symbol: Hnrnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021510.2 | NP_067485.1 | 1 - 2 | 90 | 51 | |
| GenBank mRNA | AK052983.1 | - | 1 - 2 | 44 | 51 | |
| AK088169.1 | - | 1 - 2 | 43 | 51 | ||
| BC016459.1 | - | 1 - 2 | 58 | 51 | ||
| BY787002.1 | - | 1 - 2 | 90 | 51 | ||
| Y14196.1 | - | 1 - 2 | 48 | 51 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK088169.1, NM_021510.2, Y14196.1 transcripts.
Set Membership: |
|

Hnrnph1
Genomic Map