Assay Details
| Gene Symbol: Tbc1d9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290759.1 | NP_001277688.1 | 20 - 21 | 3052 | 86 | |
| NM_001290760.1 | NP_001277689.1 | 20 - 21 | 3055 | 86 | ||
| NM_029745.2 | NP_084021.2 | 19 - 20 | 3004 | 86 | ||
| GenBank mRNA | AK035457.1 | - | 19 - 20 | 2969 | 86 | |
| AK088860.1 | - | 14 - 15 | 1954 | 86 | ||
| AK147964.1 | - | 19 - 20 | 2969 | 86 | ||
| AK172999.1 | - | 21 - 22 | 2992 | 86 | ||
| AK184661.1 | - | 3 - 4 | 145 | 86 | ||
| AK186352.1 | - | 2 - 3 | 94 | 85 | ||
| AK195900.1 | - | 3 - 4 | 145 | 86 | ||
| AK197376.1 | - | 71 | 86 | |||
| AK207874.1 | - | 2 - 3 | 94 | 86 | ||
| AK207903.1 | - | 2 - 3 | 94 | 86 | ||
| AK210511.1 | - | 2 - 3 | 94 | 86 | ||
| AK212277.1 | - | 3 - 4 | 145 | 86 | ||
| AK214532.1 | - | 2 - 3 | 94 | 86 | ||
| BC048085.1 | - | 10 - 11 | 1318 | 86 | ||
| BC058596.1 | - | 7 - 8 | 589 | 86 | ||
| BC062928.1 | - | 19 - 20 | 2991 | 6854669 | 86 | |
| BC065080.1 | - | 19 - 20 | 2983 | 6826791 | 86 | |
| BC065130.1 | - | 19 - 20 | 2947 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d9b
Genomic Map