Assay Details
| Gene Symbol: Pxdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181395.2 | NP_852060.2 | 4 - 5 | 510 | 69 | |
| GenBank mRNA | AK122223.1 | - | 4 - 5 | 280 | 69 | |
| AK142872.1 | - | 4 - 5 | 510 | 69 | ||
| AK163597.1 | - | 4 - 5 | 497 | 69 | ||
| BC150789.1 | - | 4 - 5 | 505 | 9087700 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pxdn
Genomic Map