Assay Details
| Gene Symbol: Psmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027357.2 | NP_081633.1 | 11 - 12 | 1412 | 92 | |
| GenBank mRNA | AK087947.1 | - | 11 - 12 | 1363 | 92 | |
| AK153386.1 | - | 11 - 12 | 1413 | 92 | ||
| AK159123.1 | - | 11 - 12 | 1412 | 92 | ||
| AK159750.1 | - | 11 - 12 | 1391 | 92 | ||
| AK215835.1 | - | 3 - 4 | 310 | 92 | ||
| BC022955.1 | - | 11 - 12 | 1302 | 92 | ||
| BC025802.1 | - | 11 - 12 | 1316 | 92 | ||
| BC029319.1 | - | 11 - 12 | 1349 | 92 | ||
| BC031587.1 | - | 11 - 12 | 1343 | 92 | ||
| BC032068.1 | - | 11 - 12 | 1343 | 92 | ||
| BC039937.1 | - | 11 - 12 | 1389 | 92 | ||
| BC089028.1 | - | 11 - 12 | 1359 | 92 | ||
| BC138526.1 | - | 11 - 12 | 1375 | 8861546 | 92 | |
| BC138527.1 | - | 11 - 12 | 1375 | 8861547 | 92 | |
More Information
Set Membership: |
|

Psmd1
Genomic Map