Assay Details
| Gene Symbol: Psmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027357.2 | NP_081633.1 | 12 - 13 | 1587 | 111 | |
| GenBank mRNA | AK087947.1 | - | 12 - 13 | 1538 | 111 | |
| AK153386.1 | - | 12 - 13 | 1588 | 111 | ||
| AK159123.1 | - | 12 - 13 | 1587 | 111 | ||
| AK159750.1 | - | 12 - 13 | 1566 | 111 | ||
| BC022955.1 | - | 12 - 13 | 1477 | 111 | ||
| BC025802.1 | - | 12 - 13 | 1491 | 111 | ||
| BC029319.1 | - | 12 - 13 | 1524 | 111 | ||
| BC031587.1 | - | 12 - 13 | 1518 | 111 | ||
| BC032068.1 | - | 12 - 13 | 1518 | 111 | ||
| BC039937.1 | - | 12 - 13 | 1564 | 111 | ||
| BC089028.1 | - | 12 - 13 | 1534 | 111 | ||
| BC138526.1 | - | 12 - 13 | 1550 | 8861546 | 111 | |
| BC138527.1 | - | 12 - 13 | 1550 | 8861547 | 111 | |
More Information
Set Membership: |
|

Psmd1
Genomic Map