Assay Details
| Gene Symbol: Dcaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028759.1 | NP_083035.1 | 17 - 18 | 2693 | 83 | |
| GenBank mRNA | AK004618.1 | - | 17 - 18 | 2556 | 83 | |
| AK077238.1 | - | 8 - 9 | 1211 | 83 | ||
| AK161893.1 | - | 5 - 6 | 2427 | 83 | ||
| BC058980.1 | - | 15 - 16 | 2256 | 83 | ||
| BC084732.1 | - | 15 - 16 | 2256 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dcaf6
Genomic Map