Assay Details
| Gene Symbol: Dcaf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028759.1 | NP_083035.1 | 3 - 4 | 487 | 75 | |
| GenBank mRNA | AK004618.1 | - | 3 - 4 | 350 | 75 | |
| AK215191.1 | - | 2 - 3 | 164 | 75 | ||
| BC058980.1 | - | 1 - 2 | 50 | 75 | ||
| BC084732.1 | - | 1 - 2 | 50 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf6
Genomic Map