Assay Details
| Gene Symbol: Dis3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153530.2 | NP_705758.1 | 5 - 6 | 527 | 103 | |
| GenBank mRNA | BC036177.1 | - | 5 - 6 | 488 | 5004341 | 103 |
More Information
Updated Mapping Information
Set Membership: |
|

Dis3l2
Genomic Map