Assay Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022018.3 | NP_071301.2 | 12 - 13 | 1704 | 130 | |
| GenBank mRNA | AB049355.1 | - | 12 - 13 | 1544 | 130 | |
| AK029847.1 | - | 8 - 9 | 1016 | 130 | ||
| AK136597.1 | - | 12 - 13 | 1614 | 130 | ||
| AK170347.1 | - | 3 - 4 | 299 | 130 | ||
| BC021332.1 | - | 12 - 13 | 1592 | 130 | ||
| BC038006.1 | - | 12 - 13 | 1560 | 130 | ||
| BC058234.1 | - | 10 - 11 | 1304 | 6306849 | 130 | |
| BC099690.1 | - | 12 - 13 | 1611 | 130 | ||
| BC137843.1 | - | 8 - 9 | 1081 | 8860862 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam129a
Genomic Map