| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100026403.mm.1 |
NM_025819.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026403.mm.1 |
NM_025819.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026403.mm.1 |
TC0100003253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100026405.mm.1 |
NM_025819.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026405.mm.1 |
NM_025819.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026405.mm.1 |
TC0100003253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100026402.mm.1 |
NM_025819.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026402.mm.1 |
NM_025819.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026402.mm.1 |
TC0100003253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100026404.mm.1 |
NM_025819.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026404.mm.1 |
NM_025819.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026404.mm.1 |
TC0100003253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100026406.mm.1 |
NM_025819.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026406.mm.1 |
NM_025819.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026406.mm.1 |
TC0100003253.mm.1 |
MTA TC |