Assay Details
| Gene Symbol: Swt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025819.4 | NP_080095.3 | 3 - 4 | 322 | 135 | |
| GenBank mRNA | AK004803.2 | - | 3 - 4 | 269 | 135 | |
| AK164703.1 | - | 1 - 2 | 1308 | 135 | ||
| BC028449.1 | - | 3 - 4 | 250 | 135 | ||
| BC055064.1 | - | 3 - 4 | 323 | 6528686 | 135 | |
| BC060204.1 | - | 3 - 4 | 262 | 6855442 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Swt1
Genomic Map