Assay Details
| Gene Symbol: Swt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025819.4 | NP_080095.3 | 7 - 8 | 1210 | 65 | |
| GenBank mRNA | AK004803.2 | - | 7 - 8 | 1157 | 65 | |
| BC055064.1 | - | 7 - 8 | 1211 | 6528686 | 65 | |
| BC060204.1 | - | 7 - 8 | 1150 | 6855442 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Swt1
Genomic Map