Assay Details
| Gene Symbol: Edem3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039644.2 | NP_001034733.2 | 10 - 11 | 1268 | 150 | |
| GenBank mRNA | BC060718.1 | - | 4 - 5 | 370 | 150 | |
| BC138658.1 | - | 10 - 11 | 1182 | 8861678 | 150 | |
| BC138659.1 | - | 10 - 11 | 1182 | 8861679 | 150 | |
| BC145357.1 | - | 10 - 11 | 1182 | 9053915 | 150 | |
| BC145358.1 | - | 10 - 11 | 1182 | 9053916 | 150 | |
More Information
Set Membership: |
|

Edem3
Genomic Map