Assay Details
| Gene Symbol: Coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198835.1 | NP_001185764.1 | 2 - 3 | 117 | 115 | |
| NM_007728.5 | NP_031754.1 | 1 - 2 | 316 | 115 | ||
| GenBank mRNA | AF006741.1 | - | 2 - 3 | 101 | 115 | |
| AK028690.1 | - | 1 - 2 | 203 | 115 | ||
| AK137336.1 | - | 1 - 2 | 318 | 115 | ||
| AK156430.1 | - | 2 - 3 | 86 | 115 | ||
| AK171875.1 | - | 2 - 3 | 86 | 115 | ||
| BC045137.1 | - | 2 - 3 | 68 | 6306601 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF006741.1, AK028690.1, AK137336.1, AK156430.1, AK171875.1, BC045137.1, NM_001198835.1, NM_007728.5 transcripts.
Set Membership: |
|

Coch
Genomic Map