Assay Details
| Gene Symbol: Coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198835.1 | NP_001185764.1 | 8 - 9 | 717 | 115 | |
| NM_007728.5 | NP_031754.1 | 7 - 8 | 916 | 115 | ||
| GenBank mRNA | AF006741.1 | - | 8 - 9 | 701 | 115 | |
| AK028690.1 | - | 7 - 8 | 803 | 115 | ||
| AK137336.1 | - | 7 - 8 | 918 | 115 | ||
| AK156430.1 | - | 8 - 9 | 686 | 115 | ||
| AK171875.1 | - | 8 - 9 | 686 | 115 | ||
| BC045137.1 | - | 8 - 9 | 668 | 6306601 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Coch
Genomic Map