Assay Details
| Gene Symbol: Strn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172098.1 | NP_001165569.1 | 13 - 14 | 1981 | 137 | |
| NM_052973.2 | NP_443205.1 | 15 - 16 | 2233 | 137 | ||
| GenBank mRNA | AF307777.1 | - | 15 - 16 | 2217 | 137 | |
| AK140447.1 | - | 13 - 14 | 1969 | 137 | ||
| AK165908.1 | - | 13 - 14 | 1665 | 137 | ||
| BC138055.1 | - | 13 - 14 | 1932 | 8861074 | 137 | |
| BC145127.1 | - | 13 - 14 | 1932 | 9053675 | 137 | |
| EF685152.1 | - | 16 - 17 | 2134 | 137 | ||
More Information
Set Membership: |
|

Strn3
Genomic Map