Assay Details
| Gene Symbol: Strn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172098.1 | NP_001165569.1 | 6 - 7 | 1050 | 85 | |
| NM_052973.2 | NP_443205.1 | 6 - 7 | 1050 | 85 | ||
| GenBank mRNA | AF307777.1 | - | 6 - 7 | 1034 | 85 | |
| AK139183.1 | - | 6 - 7 | 1034 | 85 | ||
| AK140447.1 | - | 6 - 7 | 1037 | 85 | ||
| BC138055.1 | - | 6 - 7 | 1001 | 8861074 | 85 | |
| BC145127.1 | - | 6 - 7 | 1001 | 9053675 | 85 | |
| EF685154.1 | - | 1 - 2 | 123 | 85 | ||
| EF685155.1 | - | 1 - 2 | 90 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Strn3
Genomic Map