Assay Details
| Gene Symbol: Marco | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010766.2 | NP_034896.1 | 15 - 16 | 1559 | 74 | |
| GenBank mRNA | AK156388.1 | - | 15 - 16 | 1233 | 74 | |
| AK159094.1 | - | 2 - 3 | 64 | 74 | ||
| AK169230.1 | - | 15 - 16 | 1235 | 74 | ||
| BC132338.1 | - | 15 - 16 | 1499 | 40130615 | 74 | |
| BC137939.1 | - | 15 - 16 | 1499 | 8860958 | 74 | |
| U18424.1 | - | 15 - 16 | 1411 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Marco
Genomic Map