Assay Details
| Gene Symbol: 2610027F03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011565.1 | - | 5 - 6 | 690 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | engrailed 1 | En1 | 
					En-1, Mo-en.1  | 
					-80.6kb | Sense | Chr. 1 - 120602391 - 120607991 on NCBI Build GRCm38 | 
| Downstream | cerebellum expressed regulatory RNA | Celrr | 
					AI848258, B230209K01Rik, Celr, Gm818  | 
					398.2kb | Sense | Chr. 1 - 121087405 - 121120975 on NCBI Build GRCm38 | 
More Information
Set Membership: | 
		   
					
						
							 | 
				

2610027F03Rik
Genomic Map