Assay Details
| Gene Symbol: Ids | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010498.3 | NP_034628.2 | 8 - 9 | 1368 | 121 | |
| GenBank mRNA | AJ627044.1 | - | 6 - 7 | 833 | 121 | |
| AK085237.1 | - | 9 - 10 | 1245 | 121 | ||
| AK132287.1 | - | 9 - 10 | 1248 | 121 | ||
| AK166178.1 | - | 8 - 9 | 1246 | 121 | ||
| AK166443.1 | - | 9 - 10 | 1197 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2128303 | CD740869.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2128303 | NM_010498.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129957 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168490 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105987 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2128303 | CD740869.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2128303 | NM_010498.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P278554 | L07921.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P278554 | AK132287.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P278554 | AK166178.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P278554 | AK166443.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P278554 | AF538870.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P173766 | L07921.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P173766 | X75636.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P173766 | AJ627044.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P438210 | AK132287.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P438210 | AK166178.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P438210 | AK166443.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P438210 | NM_010498.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P438210 | AK085237.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32448320 | - | - |
| Whole Mouse Genome G4122A | A_52_P123018 | CD740869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123018 | NM_010498.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438210 | AK132287.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438210 | AK166178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438210 | AK166443.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438210 | NM_010498.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P438210 | AK085237.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278554 | L07921.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278554 | AK132287.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278554 | AK166178.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278554 | AK166443.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P278554 | AF538870.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P173766 | L07921.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P173766 | X75636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P173766 | AJ627044.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109765 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124258 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14910238 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14910241 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14910245 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14910235 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10604908 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604909 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604907 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10604914 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421216_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427813_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455481_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434751_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450166_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427812_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10604906 | - | - |
| Mouse Genome 430 2.0 Array | 1421216_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421216_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455481_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427813_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434751_at | - | - |
| Mouse Genome 430 2.0 Array | 1450166_at | - | - |
| Mouse Genome 430 2.0 Array | 1427812_at | - | - |
| Mouse Genome 430 2.0 Array | 1450166_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455481_at | - | - |
| Mouse Genome 430 2.0 Array | 1434751_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427813_at | - | - |
| Mouse Genome 430 2.0 Array | 1427812_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15931_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15931_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00014733.mm.1 | NM_010498.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014733.mm.1 | NM_010498.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014733.mm.1 | TC0X00002384.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00014734.mm.1 | NM_010498.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014734.mm.1 | NM_010498.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014734.mm.1 | TC0X00002384.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00014736.mm.1 | NM_010498.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014736.mm.1 | NM_010498.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014736.mm.1 | TC0X00002384.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00014735.mm.1 | NM_010498.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014735.mm.1 | NM_010498.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00014735.mm.1 | TC0X00002384.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427813_at | - | - |
| Mouse Expression Set 430 | 1421216_PM_a_at | - | - |
| Mouse Expression Set 430 | 1427813_PM_at | - | - |
| Mouse Expression Set 430 | 1427812_PM_at | - | - |
| Mouse Expression Set 430 | 1434751_PM_at | - | - |
| Mouse Expression Set 430 | 1427812_at | - | - |
| Mouse Expression Set 430 | 1450166_PM_at | - | - |
| Mouse Expression Set 430 | 1450166_at | - | - |
| Mouse Expression Set 430 | 1421216_a_at | - | - |
| Mouse Expression Set 430 | 1434751_at | - | - |
| Mouse Expression Set 430 | 1455481_at | - | - |
| Mouse Expression Set 430 | 1455481_PM_at | - | - |
| Murine Genome U74v2 Set | 99883_g_at | - | - |
| Murine Genome U74v2 Set | 99882_at | - | - |
| Murine Genome U74v2 Set | 99882_PM_at | - | - |
| Murine Genome U74v2 Set | 99884_PM_at | - | - |
| Murine Genome U74v2 Set | 99884_at | - | - |
| Murine Genome U74v2 Set | 99883_PM_g_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450166_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427812_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427813_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427812_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450166_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421216_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434751_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427813_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421216_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434751_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427813_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427812_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434751_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427812_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427813_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434751_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450166_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450166_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421216_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421216_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455481_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455481_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 360619 | AK172979.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 360619 | BC033574.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 360619 | NM_145952.3 | refseq_mrna_mouse_NM |
| MouseWG-6 v2 Expression BeadChip | 5720161 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106520056 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106520056 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720161 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 360619 | AK172979.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 360619 | BC033574.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 360619 | NM_145952.3 | refseq_mrna_mouse_NM |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000360619 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2648589 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769978 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214470 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2769978 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2648589 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002947.1_141-S | - | - |
| mouse-6 v1.1 (Illumina) | scl54186.10_25-S | - | - |
| mouse-6 v1.1 (Illumina) | scl015931.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 360619 | AK172979.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 360619 | BC033574.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 360619 | NM_145952.3 | refseq_mrna_mouse_NM |
| Illumina Sentrix mouse-6.v1 bead array | 5720161 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106520056 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl015931.1_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002947.1_141-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360619 | AK172979.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360619 | BC033574.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360619 | NM_145952.3 | refseq_mrna_mouse_NM |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720161 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106520056 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2426983 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2769978 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3114518 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3114518 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214470 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2769978 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2426983 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl015931.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0002947.1_141-S | - | - |

Ids
Genomic Map