Assay Details
| Gene Symbol: Fabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024406.2 | NP_077717.1 | 2 - 3 | 310 | 73 | |
| GenBank mRNA | AK003143.1 | - | 2 - 3 | 311 | 73 | |
| AK088535.1 | - | 2 - 3 | 311 | 73 | ||
| BC054426.1 | - | 2 - 3 | 293 | 5029359 | 73 | |
| CT010390.1 | - | 2 - 3 | 245 | 73 | ||
| K02109.1 | - | 2 - 3 | 288 | 73 | ||
| M28726.1 | - | 2 - 3 | 215 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fabp4
Genomic Map