Assay Details
| Gene Symbol: Sparc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290817.1 | NP_001277746.1 | 7 - 8 | 884 | 61 | |
| NM_009242.5 | NP_033268.1 | 7 - 8 | 887 | 61 | ||
| GenBank mRNA | AK003162.1 | - | 7 - 8 | 659 | 61 | |
| AK014286.1 | - | 7 - 8 | 687 | 61 | ||
| AK049692.1 | - | 7 - 8 | 673 | 61 | ||
| AK133377.1 | - | 7 - 8 | 657 | 61 | ||
| AK146935.1 | - | 7 - 8 | 657 | 61 | ||
| AK148670.1 | - | 7 - 8 | 672 | 61 | ||
| AK159249.1 | - | 7 - 8 | 714 | 61 | ||
| AK161229.1 | - | 7 - 8 | 675 | 61 | ||
| BC004638.1 | - | 7 - 8 | 659 | 3586402 | 61 | |
| CT010306.1 | - | 6 - 7 | 581 | 61 | ||
| X04017.1 | - | 7 - 8 | 670 | 61 | ||
| X12697.1 | - | 2 - 3 | 218 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sparc
Genomic Map