Assay Details
| Gene Symbol: Serpinb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172852.3 | NP_766440.2 | 2 - 3 | 223 | 62 | |
| GenBank mRNA | AK030670.1 | - | 2 - 3 | 225 | 62 | |
| BC117990.1 | - | 2 - 3 | 213 | 40104505 | 62 | |
| BC117991.1 | - | 2 - 3 | 213 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC117991.1 transcripts.
Set Membership: |
|

Serpinb13
Genomic Map