Assay Details
| Gene Symbol: Serpinb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172852.3 | NP_766440.2 | 4 - 5 | 414 | 145 | |
| GenBank mRNA | AK030670.1 | - | 4 - 5 | 416 | 145 | |
| BC117990.1 | - | 4 - 5 | 404 | 40104505 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb13
Genomic Map