Assay Details
| Gene Symbol: Serpinb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009257.3 | NP_033283.1 | 1 - 2 | 82 | 98 | |
| GenBank mRNA | AK136532.1 | - | 1 - 2 | 88 | 98 | |
| BC005434.1 | - | 1 - 2 | 83 | 3600282 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK136532.1, BC005434.1, NM_009257.3 transcripts.
Set Membership: |
|

Serpinb5
Genomic Map