Assay Details
| Gene Symbol: Selp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011347.2 | NP_035477.1 | 11 - 12 | 2040 | 70 | |
| GenBank mRNA | AK136243.1 | - | 2 - 3 | 408 | 70 | |
| BC109158.1 | - | 10 - 11 | 1926 | 40047544 | 70 | |
| BC109159.1 | - | 10 - 11 | 1926 | 40047547 | 70 | |
| M72332.1 | - | 11 - 12 | 2040 | 70 | ||
| M87861.1 | - | 10 - 11 | 1926 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Selp
Genomic Map