Assay Details
| Gene Symbol: Selp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011347.2 | NP_035477.1 | 14 - 15 | 2375 | 67 | |
| GenBank mRNA | BC109158.1 | - | 13 - 14 | 2261 | 40047544 | 67 |
| BC109159.1 | - | 13 - 14 | 2261 | 40047547 | 67 | |
| M72332.1 | - | 14 - 15 | 2375 | 67 | ||
| M87861.1 | - | 13 - 14 | 2261 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Selp
Genomic Map