Assay Details
| Gene Symbol: Ddx59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026500.3 | NP_080776.1 | 4 - 5 | 1232 | 77 | |
| GenBank mRNA | AK004833.1 | - | 4 - 5 | 1233 | 77 | |
| AK133731.1 | - | 4 - 5 | 1194 | 77 | ||
| BC023840.1 | - | 4 - 5 | 1159 | 5344890 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx59
Genomic Map