Assay Details
| Gene Symbol: Ddx59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026500.3 | NP_080776.1 | 5 - 6 | 1476 | 99 | |
| GenBank mRNA | AK004833.1 | - | 5 - 6 | 1477 | 99 | |
| AK133731.1 | - | 5 - 6 | 1438 | 99 | ||
| BC023840.1 | - | 5 - 6 | 1403 | 5344890 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx59
Genomic Map