Assay Details
| Gene Symbol: Zswim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145456.2 | NP_663431.2 | 4 - 5 | 1322 | 152 | |
| GenBank mRNA | AK122528.1 | - | 4 - 5 | 737 | 152 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zswim6
Genomic Map