Assay Details
| Gene Symbol: Zswim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145456.2 | NP_663431.2 | 6 - 7 | 1676 | 79 | |
| GenBank mRNA | AK037945.1 | - | 2 - 3 | 271 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zswim6
Genomic Map