Assay Details
| Gene Symbol: Zswim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145456.2 | NP_663431.2 | 9 - 10 | 2241 | 71 | |
| GenBank mRNA | AK037945.1 | - | 5 - 6 | 836 | 71 | |
| AK122528.1 | - | 8 - 9 | 1509 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zswim6
Genomic Map