Assay Details
| Gene Symbol: Depdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178683.4 | NP_848798.1 | 5 - 6 | 788 | 62 | |
| GenBank mRNA | AK036550.1 | - | 5 - 6 | 747 | 62 | |
| AK043283.1 | - | 745 | 62 | |||
| AK077676.1 | - | 5 - 6 | 789 | 62 | ||
| AK083480.1 | - | 5 - 6 | 784 | 62 | ||
| BC152330.1 | - | 5 - 6 | 793 | 40141538 | 62 | |
| BC152331.1 | - | 5 - 6 | 793 | 40141540 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Depdc1b
Genomic Map