Assay Details
| Gene Symbol: Depdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178683.4 | NP_848798.1 | 6 - 7 | 843 | 66 | |
| GenBank mRNA | AK036550.1 | - | 6 - 7 | 802 | 66 | |
| AK043283.1 | - | 800 | 66 | |||
| AK077676.1 | - | 6 - 7 | 844 | 66 | ||
| AK083480.1 | - | 6 - 7 | 839 | 66 | ||
| BC152330.1 | - | 6 - 7 | 848 | 40141538 | 66 | |
| BC152331.1 | - | 6 - 7 | 848 | 40141540 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Depdc1b
Genomic Map