Assay Details
| Gene Symbol: Itgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013566.2 | NP_038594.2 | 7 - 8 | 1161 | 66 | |
| GenBank mRNA | AK078871.1 | - | 7 - 8 | 1162 | 66 | |
| AK144773.1 | - | 7 - 8 | 1134 | 66 | ||
| AK154766.1 | - | 7 - 8 | 1162 | 66 | ||
| AK155207.1 | - | 7 - 8 | 1128 | 66 | ||
| AK155594.1 | - | 7 - 8 | 1134 | 66 | ||
| BC011184.1 | - | 7 - 8 | 1134 | 4187781 | 66 | |
| M68903.1 | - | 7 - 8 | 1109 | 66 | ||
| M95632.1 | - | 7 - 8 | 1105 | 66 | ||
| M95633.1 | - | 7 - 8 | 1105 | 66 | ||
| S44607.1 | - | 6 - 7 | 1073 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgb7
Genomic Map