Assay Details
| Gene Symbol: Map3k12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163643.1 | NP_001157115.1 | 7 - 8 | 1411 | 92 | |
| NM_009582.4 | NP_033608.3 | 5 - 6 | 1235 | 92 | ||
| GenBank mRNA | AK029882.1 | - | 5 - 6 | 1044 | 92 | |
| AK034374.1 | - | 7 - 8 | 1227 | 92 | ||
| AK166435.1 | - | 7 - 8 | 1331 | 92 | ||
| BC047158.1 | - | 4 - 5 | 1029 | 6494216 | 92 | |
| BC057572.1 | - | 7 - 8 | 1235 | 6401321 | 92 | |
| U14636.1 | - | 5 - 6 | 1067 | 92 | ||
| U23789.1 | - | 4 - 5 | 1025 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k12
Genomic Map