Assay Details
| Gene Symbol: Krt6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008476.3 | NP_032502.3 | 4 - 5 | 939 | 91 | |
| GenBank mRNA | AK009394.1 | - | 4 - 5 | 942 | 91 | |
| BC064470.1 | - | 4 - 5 | 913 | 91 | ||
| BC080820.1 | - | 4 - 5 | 939 | 30141946 | 91 | |
| K02108.1 | - | 4 - 5 | 931 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | BC064470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | BC080820.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | K02108.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | NM_008476.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | AK009394.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | BC064470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | BC080820.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | K02108.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | NM_008476.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P104658 | AK009394.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218706 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362428 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163724 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156015 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | BC064470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | BC080820.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | K02108.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | NM_008476.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | AK009394.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | BC064470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | BC080820.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | K02108.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | NM_008476.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P104658 | AK009394.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | BC064470.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | BC080820.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | K02108.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | NM_008476.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | AK009394.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | BC064470.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | BC080820.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | K02108.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | NM_008476.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104658 | AK009394.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493316 | - | - |
| Whole Mouse Genome G4122A | A_52_P104658 | BC064470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104658 | BC080820.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104658 | K02108.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104658 | NM_008476.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104658 | AK009394.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104658 | BC064470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104658 | BC080820.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104658 | K02108.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104658 | NM_008476.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104658 | AK009394.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100345 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12287014 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12287008 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12286995 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12286975 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12286987 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12286983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12307753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12027420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12451681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12451680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12286981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12432850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12432847 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10432782 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432783 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432781 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432784 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422784_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422783_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427700_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432780 | - | - |
| Mouse Genome 430 2.0 Array | 1422784_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422783_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422784_at | - | - |
| Mouse Genome 430 2.0 Array | 1422783_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427700_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427700_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16687_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16687_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500018668.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018668.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018668.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018670.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018670.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018670.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018673.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018673.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018673.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018676.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018676.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018676.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018674.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018674.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018674.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018671.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018671.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018671.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018669.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018669.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018669.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018677.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018677.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018677.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018675.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018675.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018675.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018672.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018672.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018672.mm.1 | TC1500002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018667.mm.1 | NM_008476.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018667.mm.1 | NM_008476.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018667.mm.1 | TC1500002268.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422784_PM_at | - | - |
| Mouse Expression Set 430 | 1422783_PM_a_at | - | - |
| Mouse Expression Set 430 | 1422783_a_at | - | - |
| Mouse Expression Set 430 | 1427700_PM_x_at | - | - |
| Mouse Expression Set 430 | 1422784_at | - | - |
| Mouse Expression Set 430 | 1427700_x_at | - | - |
| Murine Genome U74v2 Set | 104369_at | - | - |
| Murine Genome U74v2 Set | 104369_PM_at | - | - |
| Murine Genome U74v2 Set | 104370_PM_s_at | - | - |
| Murine Genome U74v2 Set | 104370_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422783_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422784_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422783_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427700_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427700_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422784_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422783_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427700_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422784_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422784_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422783_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427700_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 476045 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6520398 | - | - |
| MouseWG-6 v2 Expression BeadChip | 2320292 | - | - |
| MouseWG-6 v2 Expression BeadChip | 510402 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6520398 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 510402 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2320292 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320292 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510402 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228295 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621685 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2738369 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2738369 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2621685 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228295 | - | - |
| mouse-6 v1.1 (Illumina) | scl016687.6_25-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0016687.2_101-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0016687.2_66-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520398 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510402 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320292 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0016687.2_66-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl016687.6_25-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0016687.2_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320292 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510402 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2738369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228295 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228295 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2738369 | - | - |

Krt6a
Genomic Map