Assay Details
| Gene Symbol: Npff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018787.1 | NP_061257.1 | 1 - 2 | 112 | 155 | |
| GenBank mRNA | AF148701.1 | - | 1 - 2 | 112 | 155 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF148701.1, NM_018787.1 transcripts.
Set Membership: |
|

Npff
Genomic Map