Assay Details
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153416.2 | NP_700465.2 | 6 - 7 | 686 | 87 | |
| GenBank mRNA | AK034591.1 | - | 6 - 7 | 581 | 87 | |
| AK079849.1 | - | 6 - 7 | 623 | 87 | ||
| AK083401.1 | - | 6 - 7 | 599 | 87 | ||
| AK083537.1 | - | 6 - 7 | 607 | 87 | ||
| AK087134.1 | - | 6 - 7 | 662 | 87 | ||
| AK088247.1 | - | 6 - 7 | 603 | 87 | ||
| AK134406.1 | - | 6 - 7 | 577 | 87 | ||
| AK165243.1 | - | 6 - 7 | 601 | 87 | ||
| AK166118.1 | - | 6 - 7 | 601 | 87 | ||
| AK169523.1 | - | 6 - 7 | 638 | 87 | ||
| AK189195.1 | - | 5 - 6 | 377 | 87 | ||
| BC018191.1 | - | 6 - 7 | 587 | 3994248 | 87 | |
| BC025501.1 | - | 6 - 7 | 633 | 5321013 | 87 | |
| BC026604.1 | - | 5 - 6 | 1140 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087134.1 transcripts.
Set Membership: |
|

Aaas
Genomic Map