Assay Details
| Gene Symbol: Krt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008475.2 | NP_032501.2 | 4 - 5 | 922 | 92 | |
| GenBank mRNA | BC064008.1 | - | 4 - 5 | 903 | 92 | |
| BC145839.1 | - | 4 - 5 | 877 | 40131142 | 92 | |
| BC145841.1 | - | 4 - 5 | 877 | 40131144 | 92 | |
| X03491.1 | - | 4 - 5 | 974 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt4
Genomic Map